(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction

Supplier: Thermo Fisher Scientific
Danger

392380025 392380010 392380100 392382500
ACRO392380025EA 457 EUR
ACRO392380025 ACRO392380010 ACRO392380100 ACRO392382500
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
(±)-2-Methyltetrahydrofuran
Formula: C₅H₁₀O
Boiling Pt: 78…80 °C (760 mmHg)
Melting Pt: –136 °C
Density: 0,86 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00005367
CAS Number: 96-47-9
EINECS: 202-507-4
UN: 2536
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.05 %
GC ≥99.0 %
Stabilizer 150 to 400 ppm BHT

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR