Diammonium hydrogen citrate 98%, pure

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Citric acid diammonium salt, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Ammonium citrate dibasic

392505000 392500025 392501000
ACRO392505000EA 66.3 EUR
ACRO392505000 ACRO392500025 ACRO392501000
Diammonium hydrogen citrate 98%, pure
di-Ammonium hydrogen citrate
Formula: HOC(CO₂H)(CH₂CO₂NH₄)₂
Density: 1,5 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00013068
CAS Number: 3012-65-5

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR