2-Bromo-α-methylbenzyl alcohol 99%

Supplier: Thermo Fisher Scientific
Warning

393640250 393640010 393640050
ACRO393640250EA 215 EUR
ACRO393640250 ACRO393640010 ACRO393640050
2-Bromo-α-methylbenzyl alcohol 99%
2-Bromo-α-methylbenzyl alcohol
Formula: C₈H₉BrO
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00065001
CAS Number: 5411-56-3
EINECS: 226-491-3

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5590 to 1.5610 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR