1-Boc-4-(hydroxymethyl)piperidine 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate, 1-Boc-4-piperidinemethanol, N-BOC-4-Piperidinemethanol

397720010 397720050
ACRO397720010EA 19.7 EUR
ACRO397720010 ACRO397720050
1-Boc-4-(hydroxymethyl)piperidine 97%
1-Boc-4-(hydroxymethyl)piperidine
Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD02094488
CAS Number: 123855-51-6

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
GC ≥96.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR