4-Fluorophenethyl bromide 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Fluoro-4-(2-bromoethyl)benzene, 1-(2-bromoethyl)-4-fluorobenzene

398050050
ACRO398050050EA 77 EUR
ACRO398050050
4-Fluorophenethyl bromide 97%
4-Fluorophenethyl bromide
Formula: C₈H₈BrF
Boiling Pt: 100…104 °C (15 mmHg)
Density: 1,4498 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD03844794
CAS Number: 332-42-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5310 to 1.5350 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR