9,9-Di-n-octylfluorene-2,7-diboronic acid 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Boronic acid,B,B'-(9,9-dioctyl-9H-fluorene-2,7-diyl)bis-, (9,9-Dioctyl-9H-fluorene-2,7-diyl)diboronic acid, 9,9-Dioctylfluorene-2,7-diboronic acid

398310050
ACRO398310050EA 248 EUR
ACRO398310050
9,9-Di-n-octylfluorene-2,7-diboronic acid 98%
9,9-Di-n-octylfluorene-2,7-diboronic acid
Formula: C₂₉H₄₄B₂O₄
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD03701608
CAS Number: 258865-48-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR