3-Fluorosalicylaldehyde

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Fluoro-2-hydroxybenzaldehyde

398890010
ACRO398890010EA 250 EUR
ACRO398890010
3-Fluorosalicylaldehyde
3-Fluorosalicylaldehyde
Formula: C₇H₅FO₂
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00003319
CAS Number: 394-50-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 67 to 72 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR