Methyl blue, pure, certified

Supplier: Thermo Fisher Scientific
Warning

401180250
ACRO401180250EA 77.7 EUR
ACRO401180250
Methyl blue, pure, certified
Methyl blue
Formula: C₃₇H₂₇N₃Na₂O₉S₃
Storage Temperature: Ambient
MDL Number: MFCD00058509
CAS Number: 28983-56-4
EINECS: 249-113-9

Order Now

Specification Test Results

Appearance Dark brown Glistening crystalline powder or crystals
Infrared spectrum Conforms
UV Lambda max 595 to 605 nm (in 0.005M HCl)
Absorption Ratio P-15/P+15 0.96 to 1.05
Titration with TiCl3 ≥12.0 ml 0.1N TiCl3/g
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR