p-Anisidinium chloride 99%

Supplier: Thermo Fisher Scientific
Danger

401300250 401301000 401300050
ACRO401300250EA 55.7 EUR
ACRO401300250 ACRO401301000 ACRO401300050
p-Anisidinium chloride 99%
p-Anisidinium chloride
Formula: C₇H₁₀ClNO
Melting Pt: 216…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00036388
CAS Number: 20265-97-8
EINECS: 243-657-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to pink to grey-brown Crystalline powder
Infrared spectrum Conforms
Melting point 214 to 219 °C
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR