2-Bromoisobutyryl bromide 98%

Supplier: Thermo Fisher Scientific
Danger

403090050 403091000 403095000
ACRO403090050EA 33.3 EUR
ACRO403090050 ACRO403091000 ACRO403095000
2-Bromoisobutyryl bromide 98%
2-Bromoisobutyryl bromide
Formula: C₄H₆Br₂O
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000122
CAS Number: 20769-85-1
EINECS: 244-017-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5070 to 1.5100 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR