Bromophenol blue ACS

Supplier: Thermo Fisher Scientific

Synonyms: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein

403140050 403140100 403140500
ACRO403140050EA 49.5 EUR
ACRO403140050 ACRO403140100 ACRO403140500
Bromophenol blue ACS
Bromophenol blue
Formula: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
Melting Pt: 279 °C
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2
Merck Index: 13,01429

Order Now

Specification Test Results

Appearance Pink-orange to red to purple Powder
Infrared spectrum Conforms
Visual transition interval from pH 3.0 (yellow) to pH 4.6 (blue)
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR