3-Bromo-1-propanol 92%

Supplier: Thermo Fisher Scientific
Warning

403200250 403200050
ACRO403200250EA 127 EUR
ACRO403200250 ACRO403200050
3-Bromo-1-propanol 92%
3-Bromo-1-propanol
Formula: BrH₂CCH₂CH₂OH
Boiling Pt: 62 °C (5 mmHg)
Density: 1,6 g/cm³
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002942
CAS Number: 627-18-9
EINECS: 210-986-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4850 to 1.4900 (20°C, 589 nm)
GC ≥90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR