2-Bromopyrimidine 97+%

Supplier: Thermo Fisher Scientific
Warning

403230010 403230050
ACRO403230010EA 34.5 EUR
ACRO403230010 ACRO403230050
2-Bromopyrimidine 97+%
2-Bromopyrimidine
Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Boiling Pt: 62…64 °C (1.5 mmHg)
Melting Pt: 55…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00014601
CAS Number: 4595-60-2

Order Now

Specification Test Results

Appearance Off-white Crystalline powder
Infrared spectrum Conforms
Melting point 55 to 59 °C
HPLC ≥97.0 %
Specific optical rotation -11° to -16° (20 °C, 589 nm) (c=4, MeOH)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR