Charcoal activated, NORIT® SA 2, decolorizing

Supplier: Thermo Fisher Scientific

404035000 404030025
ACRO404035000EA 65.7 EUR
ACRO404035000 ACRO404030025
Charcoal activated, NORIT® SA 2, decolorizing
Charcoal activated
Formula: C
Melting Pt: 3652 °C
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Specification Test Results

Appearance Black Powder
Loss on drying ≤10 %
Molasses number ≤390
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR