4-Chlorophenylhydrazinium chloride 97%

Supplier: Thermo Fisher Scientific
Warning

404950250 404950050 404951000
ACRO404950250EA 116 EUR
ACRO404950250 ACRO404950050 ACRO404951000
4-Chlorophenylhydrazinium chloride 97%
4-Chlorophenylhydrazinium chloride
Formula: C₆H₈Cl₂N₂
Melting Pt: 203 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012943
CAS Number: 1073-70-7
EINECS: 214-030-9

Order Now

Specification Test Results

Appearance White to pink to light brown Crystalline powder
Infrared spectrum Conforms
Titration Argentometric 96.0 to 104.0 %
Loss on drying ≤3 % (1 g, 105°C)
HPLC ≥96.0 %
Residue after evaporation ≤0.0005 %
Free amine ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR