Diethyl chlorophosphate 95%

Supplier: Thermo Fisher Scientific
Danger

407241000 407240250 407245000
ACRO407241000EA 122 EUR
ACRO407241000 ACRO407240250 ACRO407245000
Diethyl chlorophosphate 95%
Diethyl chlorophosphate
Formula: C₄H₁₀ClO₃P
MW: 172.55 g/mol
Boiling Pt: 81 °C (6 mmHg)
Density: 1.19 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00009075
CAS Number: 814-49-3
EINECS: 212-396-4
UN: 2927
ADR: 6.1 I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4160 to 1.4180 (20°C, 589 nm)
GC ≥94 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR