Fluorescein disodium salt dihydrate 90%, pure

Supplier: Thermo Fisher Scientific

410641000 410645000
ACRO410641000EA 41 EUR
ACRO410641000 ACRO410645000
Fluorescein disodium salt dihydrate 90%, pure
Fluorescein disodium salt dihydrate

CAS No.: 1264198-62-0

Formula: C₂₀H₁₀Na₂O₅·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150814
CAS Number: 1264198-62-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Orange-red to green-brown to dark brown Powder
Infrared spectrum Conforms
Water 6.5 to 10 % (K.F.)
UV spectrophotometry ≥88 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR