L(+)-Histidine monohydrochloride monohydrate 98%

Supplier: Thermo Fisher Scientific

Synonyms: L-Histidine monohydrochloride monohydrate, (2S)-(+)-Histidine monohydrochloride monohydrate, L-Histidine hydrochloride monohydrate, (S)-(+)-Histidine monohydrochloride monohydrate, His HCl monohydrate, H-His-OH.HCl.H2O, H-His-OH·HCl·H₂O, L-3-Imidazol-4-ylalanine monohydrochloride monohydrate, L-Histidine hydrochloride hydrate

411730250 411731000 411730025 411735000
ACRO411730250EA 50.7 EUR
ACRO411730250 ACRO411731000 ACRO411730025 ACRO411735000
L(+)-Histidine monohydrochloride monohydrate 98%
L(+)-Histidine monohydrochloride monohydrate
Formula: C₆H₉N₃O₂·HCl·H₂O
MW: 209.63 g/mol
Melting Pt: 259
Storage Temperature: Ambient
MDL Number: MFCD00151027
CAS Number: 5934-29-2
EINECS: 211-438-9
Merck Index: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561d2381f930afe618f78d680790ea9a43f6596c&vid=8aacf6dc4ef5382410cb743498272b48c3c36999&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying 7 % to 10 % (3 h) (145 to 150°C)
Sulfated ash ≤0.1 %
pH 3 to 5 (2.5 % at 25°C)
Specific optical rotation +9.2° to +10.6° (20°C, 589 nm) (c=11, 6 N HCl) (On anhydrous substance)
Titration with NaOH ≥97.5 % (On anhydrous substance)
Sulfate (SO4) ≤300 ppm
Ammonium (NH4) ≤200 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Other amino acids ≤0.5 % (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR