Pyranine 98%

Supplier: Thermo Fisher Scientific
Warning

412180050 412180010 412180250
ACRO412180050EA 133 EUR
ACRO412180050 ACRO412180010 ACRO412180250
Pyranine 98%
Pyranine

CAS No.: 6358-69-6

Formula: C₁₆H₇Na₃O₁₀S₃
Density: 2,15 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00037575
CAS Number: 6358-69-6

Order Now

Specification Test Results

Appearance Yellow to yellow-green Powder or crystals
Infrared spectrum Conforms
UV spectrophotometry ≥97.5 %
UV Lambda max 450 to 458 nm (in 0.01N NaOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR