4-Iodobenzonitrile 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: p-Iodobenzonitrile

412380250 412380050
ACRO412380250EA 263 EUR
ACRO412380250 ACRO412380050
4-Iodobenzonitrile 99%
4-Iodobenzonitrile
Formula: C₇H₄IN
MW: 229.02 g/mol
Boiling Pt: 285
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00051310
CAS Number: 3058-39-7
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Light yellow to yellow-brown Powder and/or chunks
Infrared spectrum Conforms
Melting point 125 to 128 °C
GC ≥98.5 %
Stabilizer 50 to 150 ppm Amylene

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR