Indocyanine green, pure laser grade

Supplier: Thermo Fisher Scientific
Warning

412541000 412545000 412540020
ACRO412541000EA 125 EUR
ACRO412541000 ACRO412545000 ACRO412540020
Indocyanine green, pure laser grade
Indocyanine green
Formula: C₄₃H₄₇N₂NaO₆S₂
Melting Pt: 235 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013078
CAS Number: 3599-32-4

Order Now

Specification Test Results

Appearance Green to brown Crystalline powder or crystals
Infrared spectrum Conforms
UV Lambda max 784 nm (in Methanol)
Molar absorption coefficient ≥230000 l/(mol.cm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR