Nickel(II) phthalocyanine 95%

Supplier: Thermo Fisher Scientific
Danger

415600050 415600250
ACRO415600050EA 61.9 EUR
ACRO415600050 ACRO415600250
Nickel(II) phthalocyanine 95%
Nickel(II) phthalocyanine

Appearance: Dark blue Crystalline powder

Formula: Ni(C₃₂H₁₆N₈)
Storage Temperature: Ambient
MDL Number: MFCD00041954
CAS Number: 14055-02-8

Order Now

Specification Test Results

Appearance Dark blue Crystalline powder
Infrared spectrum Conforms
Gravimetry ≥94 %
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR