4-Nitrophthalic acid 99%

Supplier: Thermo Fisher Scientific
Warning

416120250 416121000
ACRO416120250EA 233 EUR
ACRO416120250 ACRO416121000
4-Nitrophthalic acid 99%
4-Nitrophthalic acid
Formula: C₈H₅NO₆
MW: 211.13 g/mol
Melting Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00007252
CAS Number: 610-27-5
EINECS: 210-215-3

Order Now

Specification Test Results

Appearance Yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 163 to 167 °C
HPLC ≥98.5 %
Titration with TBAH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR