Pinacol 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2,3-Dimethylbutane-2,3-diol, Tetramethylethylene glycol, 2,3-Dimethyl-2,3-butanediol

418001000 418002500 418000010
ACRO418001000EA 159 EUR
ACRO418001000 ACRO418002500 ACRO418000010
Pinacol 99%
Pinacol
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 174 °C
Melting Pt: 40…43 °C
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00004462
CAS Number: 76-09-5
EINECS: 200-933-5
Merck Index: 13,07521

Order Now

Specification Test Results

Appearance White Low melting crystalline solid
Infrared spectrum Conforms
GC ≥98.5 %
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR