Potassium chlorate 99+% ACS

Supplier: Thermo Fisher Scientific
Danger

418190050 418195000 418190025
ACRO418190050EA 29.8 EUR
ACRO418190050 ACRO418195000 ACRO418190025
Potassium chlorate 99+% ACS
Potassium chlorate
Formula: KClO₃
Boiling Pt: 400 °C (1013 hPa)
Melting Pt: 356…368 °C
Density: 2,34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011361
CAS Number: 3811-04-9
EINECS: 223-289-7
UN: 1485
ADR: 5.1,II
Merck Index: 13,07703

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration ≥99.0 %
Chloride (Cl) ≤10 ppm
Bromate (BrO3) ≤150 ppm
Sulfate (SO4) Passes test (limit about 0.002%)
Calcium (Ca) ≤20 ppm
Iron (Fe) ≤3 ppm
Magnesium (Mg) ≤20 ppm
Sodium (Na) ≤100 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR