(S)-(+)-Prolinol 95%

Supplier: Thermo Fisher Scientific
Warning

418320010 418320050 418320250
ACRO418320010EA 35.4 EUR
ACRO418320010 ACRO418320050 ACRO418320250
(S)-(+)-Prolinol 95%
(S)-(+)-Prolinol
Formula: C₅H₁₁NO
Boiling Pt: 74…76 °C (2 mmHg)
Density: 1,036 g/cm³
Flash Pt: 86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005255
CAS Number: 23356-96-9
EINECS: 245-605-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4840 to 1.4870 (20°C, 589 nm)
GC ≥94 %
Specific optical rotation +29° to +33° (20°C, 589 nm) (c=1, toluene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR