Silver phosphate 98%

Supplier: Thermo Fisher Scientific
Warning

419370250
ACRO419370250EA 133 EUR
ACRO419370250
Silver phosphate 98%
Silver phosphate

Appearance: Yellow to yellow-green to dark yellow Powder

Formula: Ag₃O₄P
MW: 418.58 g/mol
Melting Pt: 849 °C
Storage Temperature: Ambient
MDL Number: MFCD00014148
CAS Number: 7784-09-0

Order Now

Specification Test Results

Appearance Yellow to yellow-green to dark yellow Powder
Assay ≥97.5 %
Particle size ≤5.0 % (> 1180 micron)
Harmonic mean size 0.600 to 0.750 mm
Uniformity coefficient ≤1.60
Moisture holding capacity 49.0 to 60.0 % (Cl)
Total volume capacity ≥1.20 eq/l (Cl)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR