Tetrabromophenolphthalein ethyl ester, pure

Supplier: Thermo Fisher Scientific

420070010 420070050 420070250
ACRO420070010EA 238 EUR
ACRO420070010 ACRO420070050 ACRO420070250
Tetrabromophenolphthalein ethyl ester, pure
Tetrabromophenolphthalein ethyl ester

CAS No.: 1176-74-5

Formula: C₂₂H₁₄Br₄O₄
Melting Pt: 208…211 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066387
CAS Number: 1176-74-5

Order Now

Specification Test Results

Appearance Yellow to red Powder
Infrared spectrum Conforms
UV spectrophotometry ≥80.0 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR