Tetramethylammonium acetate hydrate 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Tetramethylammonium acetate xhydrate

420490250 420490050
ACRO420490250EA 207 EUR
ACRO420490250 ACRO420490050
Tetramethylammonium acetate hydrate 95%
Tetramethylammonium acetate hydrate

Appearance: White Crystalline solid

Formula: C₆H₁₅NO₂·xH₂O
MW: 133.20 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD03066126
CAS Number: 1266615-86-4
EINECS: 234-179-3

Order Now

Specification Test Results

Appearance White Crystalline solid
Infrared spectrum Conforms
Water ≤14 % (K.F.)
Titration with HClO4 ≥94 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR