Tetrapropylammonium chloride 94%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Tetra-n-propylammonium chloride

420710250 420711000
ACRO420710250EA 137 EUR
ACRO420710250 ACRO420711000
Tetrapropylammonium chloride 94%
Tetrapropylammonium chloride
Formula: C₁₂H₂₈ClN
Melting Pt: 240…242 °C
Density: 1,0334 g/cm³ (3 °C)
Storage Temperature: Ambient
MDL Number: MFCD00038729
CAS Number: 5810-42-4
EINECS: 227-375-5

Order Now

Specification Test Results

Appearance White to light yellow Crystals or crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥92 %
Specific optical rotation +33° to +36° (20°C, 589 nm) (c=10, 2N HCl)
Titration with HClO4 ≥98.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR