Benzyl mercaptan 99%

Supplier: Thermo Fisher Scientific
Danger

421261000 421265000
ACRO421261000EA 57.3 EUR
ACRO421261000 ACRO421265000
Benzyl mercaptan 99%
Benzyl mercaptan
Formula: C₇H₈S
Boiling Pt: 195 °C
Melting Pt: –29 °C
Density: 1,058 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00004867
CAS Number: 100-53-8
EINECS: 202-862-5
UN: 2810
ADR: 6.1,II
Merck Index: 12,09456

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5735 to 1.5775 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR