Tridodecylamine (hydrogen ionophore I) 90%

Supplier: Thermo Fisher Scientific
Warning

421620250 421621000
ACRO421620250EA 84.8 EUR
ACRO421620250 ACRO421621000
Tridodecylamine (hydrogen ionophore I) 90%
Tridodecylamine (Hydrogen ionophore I)
Formula: C₃₆H₇₅N
Boiling Pt: 450 °C (1013 hPa)
Melting Pt: 16 °C
Density: 0,82 g/cm³
Flash Pt: 216 °C
Storage Temperature: Ambient
MDL Number: MFCD00008971
CAS Number: 102-87-4
EINECS: 203-063-4
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4610 (20°C, 589 nm)
Titration with HClO4 ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR