Tris(2-methylphenyl)phosphine 99%

Supplier: Thermo Fisher Scientific
Warning

422320050 422320010 422320250
ACRO422320050EA 202 EUR
ACRO422320050 ACRO422320010 ACRO422320250
Tris(2-methylphenyl)phosphine 99%
Tris(2-methylphenyl)phosphine
Formula: C₂₁H₂₁P
Boiling Pt: 187 - 192 °C (0,05 -rr)
Melting Pt: 126…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00008514
CAS Number: 6163-58-2
EINECS: 228-193-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 122 to 126 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR