Diammonium hydrogen orthophosphate 98+% ACS

Supplier: Thermo Fisher Scientific

423375000 423370025
ACRO423375000EA 58.9 EUR
ACRO423375000 ACRO423370025
Diammonium hydrogen orthophosphate 98+% ACS
di-Ammonium hydrogen orthophosphate
Formula: (NH₄)₂HPO₄
Melting Pt: 155 °C
Density: 1,619 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 13,00546

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
pH 7.7 to 8.1 (5 % at 25.0°C)
Titration with HCl ≥98.0 %
Chloride (Cl) ≤0.001 %
Nitrate (NO3) ≤0.003 %
Sulfate (SO4) ≤0.01 %
Calcium (Ca) ≤0.001 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.0005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR