Potassium hexacyanoferrate(III) 99+% ACS

Supplier: Thermo Fisher Scientific

Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide

424120050 424125000 424120025
ACRO424120050EA 34.2 EUR
ACRO424120050 ACRO424125000 ACRO424120025
Potassium hexacyanoferrate(III) 99+% ACS
Potassium hexacyanoferrate (III)
Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3
Merck Index: 13,07712

Order Now

Specification Test Results

Appearance Orange to red Crystals and/or chunks
Titration Iodometric ≥99.0 %
Chloride (Cl) ≤0.01 %
Sulfate (SO4) ≤0.01 %
Ferro compounds ≤0.05 % as ferrocyanide ion ¢Fe(CN)6!4-
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR