Benzyl-4-oxo-1-piperidinecarboxylate 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Cbz-Piperidin-4-one, 1-Carbobenzoxy-4-piperidone, 1-Cbz-4-piperidone, 4-Oxo-1-piperidinecarboxylic Acid Benzyl Ester, 1-Benzyloxycarbonyl-4-piperidine

425960050 425960250
ACRO425960050EA 38.4 EUR
ACRO425960050 ACRO425960250
Benzyl-4-oxo-1-piperidinecarboxylate 97%
Benzyl-4-oxo-1-piperidinecarboxylate
Formula: C₁₃H₁₅NO₃
MW: 233.27 g/mol
Boiling Pt: 114…140 °C (.3 mmHg)
Density: 1.185 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00673144
CAS Number: 19099-93-5

Order Now

Specification Test Results

Appearance Colorless to white to light yellow to light orange Powder or solid or liquid
Infrared spectrum Conforms
GC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR