Hydrogen peroxide 3% w/w water Stabilised

Supplier: Thermo Fisher Scientific

426000010 426000250 426001000
ACRO426000010EA 159 EUR
ACRO426000010 ACRO426000250 ACRO426001000
Hydrogen peroxide 3% w/w water Stabilised
Hydrogen peroxide
Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: 100 °C (760 mmHg)
Melting Pt: -0.41
Density: 1.01 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
Merck Index: 14,04798

Order Now

Specification Test Results

Appearance Clear colorless Solution
Stabilizer Approx. 300 ppm
Titration with KMnO4 3 to 4 %
GC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR