Dithiothreitol (DTT, Cleland's reagent) 1 M in water for biochemistry

Supplier: Thermo Fisher Scientific
Warning

426380100 426380500 426382500
ACRO426380100EA 39.7 EUR
ACRO426380100 ACRO426380500 ACRO426382500
Dithiothreitol (DTT, Cleland's reagent) 1 M in water for biochemistry
Dithiothreitol (DTT, Cleland's reagent)
Formula: C₄H₁₀O₂S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
UN: 3334
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration Iodimetric 0.90 to 1.10 M
UV at 500 nm A: ≤0.05

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR