4-Methoxy-2-methylbenzaldehyde 97%

Supplier: Thermo Fisher Scientific
Warning

427020010 427020050
ACRO427020010EA 52.7 EUR
ACRO427020010 ACRO427020050
4-Methoxy-2-methylbenzaldehyde 97%
4-Methoxy-2-methylbenzaldehyde
Formula: C₉H₁₀O₂
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD02261771
CAS Number: 52289-54-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR