4-Fluoro-2-(trifluoromethyl)phenylboronic acid 97%

Supplier: Thermo Fisher Scientific
Warning

430440010 430440050
ACRO430440010EA 97.2 EUR
ACRO430440010 ACRO430440050
4-Fluoro-2-(trifluoromethyl)phenylboronic acid 97%
4-Fluoro-2-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BF₄O₂
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD08062379
CAS Number: 182344-16-7

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 182 to 187 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR