4-Amino-2-hydroxypyridine 97%

Supplier: Thermo Fisher Scientific
Danger

432080010 432080050
ACRO432080010EA 170 EUR
ACRO432080010 ACRO432080050
4-Amino-2-hydroxypyridine 97%
4-Amino-2-hydroxypyridine
Formula: C₅H₆N₂O
Melting Pt: 206,5…208.5 °C
Storage Temperature: Ambient
MDL Number: MFCD02682913
CAS Number: 38767-72-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to green to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥99.7 %
UV at 245 nm A: ≤0.046
Color scale ≤10 APHA
Residue after evaporation ≤0.0003 %
Acidity ≤0.0002 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR