2-Amino-5-bromo-4-(trifluoromethyl)pyridine 97%

Supplier: Thermo Fisher Scientific
Warning

432500050
ACRO432500050EA 737 EUR
ACRO432500050
2-Amino-5-bromo-4-(trifluoromethyl)pyridine 97%
2-Amino-5-bromo-4-(trifluoromethyl)pyridine
Formula: C₆H₄BrF₃N₂
Melting Pt: 71…74 °C
Storage Temperature: Ambient
MDL Number: MFCD09864896
CAS Number: 944401-56-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
Melting point 71 to 75 °C
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR