Trifluoroacetic acid 99+% for spectroscopy

Supplier: Thermo Fisher Scientific
Danger

Synonyms: TFA

434161000
ACRO434161000EA 205 EUR
ACRO434161000
Trifluoroacetic acid 99+% for spectroscopy
Trifluoroacetic acid
Formula: C₂HF₃O₂
Boiling Pt: 72 °C (760 mmHg)
Melting Pt: –15 °C
Density: 1,535 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
UN: 2699
ADR: 8,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 %
UV at 260 nm A: ≤1.0
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR