5-Iodopyridin-2-ol 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Hydroxy-5-iodopyridine, 5-Iodopyridin-2(1H)-one

436370010
ACRO436370010EA 0 EUR
ACRO436370010
5-Iodopyridin-2-ol 97%
5-Iodopyridin-2-ol
Formula: C₅H₄INO
Melting Pt: 186,7…190.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00234058
CAS Number: 13472-79-2

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 186 to 191 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR