Ytterbium(III) chloride hexahydrate 99.99+% (trace metals basis)

Supplier: Thermo Fisher Scientific
Warning

437180500 437180100
ACRO437180500EA 373 EUR
ACRO437180500 ACRO437180100
Ytterbium(III) chloride hexahydrate 99.99+% (trace metals basis)
Ytterbium (III) chloride hexahydrate
Formula: Cl₃Yb·6H₂O
Melting Pt: 865 °C
Density: 2,57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149885
CAS Number: 10035-01-5
EINECS: 233-800-5

Order Now

Specification Test Results

Appearance White Powder or crystals
Total trace metal impurities ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR