Ethyl-α-bromophenylacetate 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethyl alpha-bromophenylacetate

438060050 438060250 438061000
ACRO438060050EA 43.9 EUR
ACRO438060050 ACRO438060250 ACRO438061000
Ethyl-α-bromophenylacetate 97%
Ethyl α-bromophenylacetate
Formula: C₁₀H₁₁BrO₂
MW: 243.10 g/mol
Boiling Pt: 89…92 °C (1.2hPa)
Density: 1.389 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013536
CAS Number: 2882-19-1
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5370 to 1.5420 (20°C, 589 nm)
GC ≥96.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR