2-Methylpropane-1-sulphonyl chloride 97%

Supplier: Thermo Fisher Scientific
Danger

438410010 438410050
ACRO438410010EA 77.4 EUR
ACRO438410010 ACRO438410050
2-Methylpropane-1-sulphonyl chloride 97%
2-Methylpropane-1-sulphonyl chloride
Formula: C₄H₉ClO₂S
Boiling Pt: 77…78 °C (11Torr)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD07369800
CAS Number: 35432-36-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR