Boc-L-alaninal 98%

Supplier: Thermo Fisher Scientific

Synonyms: N-tert-Butoxycarbonyl-L-alaninal, Boc-L-Ala-aldehyde, Boc-L-alaninal, Boc-Ala-aldehyde, (S)-(1-Oxopropan-2-yl)carbamic acid tert-Butyl ester, (S)-tert-Butyl (1-Oxopropan-2-yl)carbamate

438470010
ACRO438470010EA 235 EUR
ACRO438470010
Boc-L-alaninal 98%
Boc-L-alaninal
Formula: C₈H₁₅NO₃
Melting Pt: 86,4…87.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00143786
CAS Number: 79069-50-4

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
GC ≥97.5 %
Specific optical rotation -29° to -39° (20°C, 589 nm) (C=2 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR