5,5-Dibromopyrimidine-2,4,6(1H,3H,5H)-trione 97%

Supplier: Thermo Fisher Scientific
Danger

439840010 439840050
ACRO439840010EA 53.3 EUR
ACRO439840010 ACRO439840050
5,5-Dibromopyrimidine-2,4,6(1H,3H,5H)-trione 97%
5,5-Dibromopyrimidine-2,4,6(1H,3H,5H)-trione
Formula: C₄H₂Br₂N₂O₃
Melting Pt: 240…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00010595
CAS Number: 511-67-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR