4-Cyanopiperidine 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Piperidine-4-carbonitrile

441820050 441820250
ACRO441820050EA 64.4 EUR
ACRO441820050 ACRO441820250
4-Cyanopiperidine 97%
4-Cyanopiperidine
Formula: C₆H₁₀N₂
MW: 110.16 g/mol
Boiling Pt: 100…105 °C (12 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD05022468
CAS Number: 4395-98-6
UN: 3276
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR